Learn More
MSC Tesa Brown and Sharpe Flexible Steel Rule, 6 in., 4R Graduation, 1/2 in. Wide, 1 ea.
Catalog No. 5000514931
Encompass
This flexible steel rule is designed for precise measurements in various shop and laboratory environments. It is manufactured from hardened and tempered steel for durability and features a satin chrome finish to minimize light reflection and ensure readability.
  • Graduation style is 4R, featuring 8ths, 16ths, 32nds, and 64ths
  • Flexible construction allows for use on curved or uneven surfaces
  • Satin chrome finish resists corrosion and reduces glare
  • Dimensions are 0.5 in. wide and 6 in. overall length
  • Hardened and tempered steel for long-term reliability
Catalog No. 50-005-14931 Supplier MSC Supplier No. 06511612
This flexible steel rule is designed for precise measurements in various shop and laboratory environments. It is manufactured from hardened and tempered steel for durability and features a satin chrome finish to minimize light reflection and ensure readability.
  • Graduation style is 4R, featuring 8ths, 16ths, 32nds, and 64ths
  • Flexible construction allows for use on curved or uneven surfaces
  • Satin chrome finish resists corrosion and reduces glare
  • Dimensions are 0.5 in. wide and 6 in. overall length
  • Hardened and tempered steel for long-term reliability