Learn More
MSC PEC Tools Flexible Steel Rule, 24 in., 4R Graduation, 1 ea
Catalog No. 5000516807
Encompass
This 24 in. flexible steel rule is designed for precision measurements, featuring a durable, hardened and tempered steel construction. It includes a satin-chrome finish to reduce glare and ensure long-lasting performance in various industrial or laboratory environments.
  • Flexible body construction for versatile use
  • Satin-chrome finish provides enhanced readability
  • Graduations include 8ths, 16ths, 32nds and 64ths
  • Precisely etched and ink-filled markings for accuracy
  • Fully hardened and tempered blade
  • Dimensions measure 24 in. long by 0.75 in. wide with a thickness of 0.020 in.
  • Made in the U.S.A.
Catalog No. 50-005-16807 Supplier MSC Supplier No. 06511240
This 24 in. flexible steel rule is designed for precision measurements, featuring a durable, hardened and tempered steel construction. It includes a satin-chrome finish to reduce glare and ensure long-lasting performance in various industrial or laboratory environments.
  • Flexible body construction for versatile use
  • Satin-chrome finish provides enhanced readability
  • Graduations include 8ths, 16ths, 32nds and 64ths
  • Precisely etched and ink-filled markings for accuracy
  • Fully hardened and tempered blade
  • Dimensions measure 24 in. long by 0.75 in. wide with a thickness of 0.020 in.
  • Made in the U.S.A.