Learn More
Grainger Satin Steel Rule 1mm/.5mm
Catalog No. 502765743
Encompass_Preferred
The Mitutoyo Satin Steel Rule is a 12-inch stainless steel ruler with satin chrome finish for durability. It has clear, easy-to-read markings in 1/64" and 1 mm increments on both sides for precise measurements. The ruler is 0.5mm thick for strength and rigidity. It features a beveled edge to allow flush contact with surfaces. The stainless steel construction resists corrosion and provides chemical resistance. This high-quality ruler is made in Japan and ideal for engineers, machinists, woodworkers, and other professionals requiring accurate measurements.
Catalog No. 50-276-5743 Supplier Grainger Supplier No. 16X226
Add to Cart
Add to Cart
The Mitutoyo Satin Steel Rule is a 12-inch stainless steel ruler with satin chrome finish for durability. It has clear, easy-to-read markings in 1/64" and 1 mm increments on both sides for precise measurements. The ruler is 0.5mm thick for strength and rigidity. It features a beveled edge to allow flush contact with surfaces. The stainless steel construction resists corrosion and provides chemical resistance. This high-quality ruler is made in Japan and ideal for engineers, machinists, woodworkers, and other professionals requiring accurate measurements.