Learn More
Medchemexpress LLC Satraplatin | 129580-63-8 | MFCD00871429 | >98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1757610MG
Encompass_Preferred
Satraplatin is an orally active platinum(IV) antineoplastic research compound that acts as a DNA alkylator, forming DNA adducts that inhibit DNA replication and induce cytotoxicity. It is commonly used in oncology research to investigate platinum-based chemotherapy mechanisms and models of resistance.
  • Forms DNA adducts and induces DNA damage.
  • Useful for studies of platinum-based chemotherapy and resistance mechanisms.
  • High-purity material suitable for in vitro and in vivo research.
  • Consult the safety data sheet for handling, storage, and disposal instructions.
Catalog No. 50-002-98704