Learn More
Cayman Chemical SaturATdMonunsaturATd Fat 1ea
SDP
Catalog No. 502966848
Encompass_Preferred
Contains various saturated or unsaturated fatty acids; designed for direct use in LC-MS applications. EAch ampule contains at least 1 ml: Arachidic Acid (Item No. 9000339) Lauric Acid (Item No. 10006626) Lignoceric Acid (Item No. 13353) Myristic Acid (Item No. 13351 Nervonic Acid (Item No. 13940) Oleic Acid (Item No. 90260) Palmitic Acid (Item No. 10006627) Palmitoleic Acid (Item No. 10009871) Stearic Acid (Item No. 10011298)
Catalog No. 50-296-6848 Supplier Cayman Chemical Supplier No. 179421 EA
Contains various saturated or unsaturated fatty acids; designed for direct use in LC-MS applications. EAch ampule contains at least 1 ml: Arachidic Acid (Item No. 9000339) Lauric Acid (Item No. 10006626) Lignoceric Acid (Item No. 13353) Myristic Acid (Item No. 13351 Nervonic Acid (Item No. 13940) Oleic Acid (Item No. 90260) Palmitic Acid (Item No. 10006627) Palmitoleic Acid (Item No. 10009871) Stearic Acid (Item No. 10011298)