Learn More
Teknova Saturated Ammonium Sulfate Solution. 1000mL, Sterile.
SDP
Catalog No. 502387965
Encompass
Ammonium sulfate is a powerful inorganic disinfectant tbat can be used as a precipitating agent for both proteins and plasmids.
Catalog No. 50-238-7965 Supplier Teknova Supplier No. A2030
Ammonium sulfate is a powerful inorganic disinfectant tbat can be used as a precipitating agent for both proteins and plasmids.