Learn More
VWR INTERNATIONAL LLC SATURATED PHENOL 4.5 400MLSAT
Catalog No. NC9179446
Encompass
An ideal replacement for water-saturated phenol in many protocols. Offers the high purity and performance of water saturated phenol with greatly enhanced stability and safety.
Catalog No. NC9179446 Supplier VWR INTERNATIONAL LLC Supplier No. 97064716
An ideal replacement for water-saturated phenol in many protocols. Offers the high purity and performance of water saturated phenol with greatly enhanced stability and safety.