Learn More
INTACT GENOMICS INC Sau DNA Polymerase Large Fragment 1000 Units (5 units/µl)
Catalog No. 5000082817
Encompass
Sau DNA Polymerase I, Large Fragment is a product of the Staphylococcus aureus DNA polymerase I which lacks the N-terminal exonuclease domain (1-293 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity.Applications Strand displacement DNA synthesis (1) Random primer labeling Second strand cDNA synthesis dA-tailing
Catalog No. 50-000-82817 Supplier INTACT GENOMICS INC Supplier No. 3595
Sau DNA Polymerase I, Large Fragment is a product of the Staphylococcus aureus DNA polymerase I which lacks the N-terminal exonuclease domain (1-293 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity.Applications Strand displacement DNA synthesis (1) Random primer labeling Second strand cDNA synthesis dA-tailing