Learn More
New England Biolabs, Inc. Sau3AI – 1000 units
SDP
Catalog No. 50812076
Encompass
A restriction endonuclease that recognizes the sequence ^GATC_.
Catalog No. 50-812-076 Supplier New England Biolabs, Inc. Supplier No. R0169L
A restriction endonuclease that recognizes the sequence ^GATC_.