Learn More
New England Biolabs, Inc. Sau3AI – 200 units
SDP
Catalog No. 50812078
Encompass
A restriction endonuclease that recognizes the sequence ^GATC_.
Catalog No. 50-812-078 Supplier New England Biolabs, Inc. Supplier No. R0169S
A restriction endonuclease that recognizes the sequence ^GATC_.