Learn More
New England Biolabs, Inc. Sau96I – 1000 units
SDP
Catalog No. 50812071
Encompass
A restriction endonuclease that recognizes the sequence G^GNC_C.
Catalog No. 50-812-071 Supplier New England Biolabs, Inc. Supplier No. R0165S
A restriction endonuclease that recognizes the sequence G^GNC_C.