Learn More
Medchemexpress LLC Sauchinone | 177931-17-8 | MFCD08702698 | 356.37 | 20 MG
SDP
Catalog No. 5000388757
Encompass_Preferred
Sauchinone is a natural product isolated from *Saururus chinensis*. It is a solid compound used as a laboratory chemical and for the manufacture of substances, including applications in pharmaceutical, nutraceutical, and cosmetic research.
  • Solid appearance
  • Melting/freezing point of 224-226°C
  • Relative density of 1.43 g/cm³
  • Stable under recommended storage conditions
  • Suitable for various research applications
Catalog No. 50-003-88757 Supplier Medchemexpress LLC Supplier No. HYN061320MG
Sauchinone is a natural product isolated from *Saururus chinensis*. It is a solid compound used as a laboratory chemical and for the manufacture of substances, including applications in pharmaceutical, nutraceutical, and cosmetic research.
  • Solid appearance
  • Melting/freezing point of 224-226°C
  • Relative density of 1.43 g/cm³
  • Stable under recommended storage conditions
  • Suitable for various research applications