Learn More
AdipoGen Sauchinone (1 mg)
SDP
Supplier:  AdipoGen AGCN20032M001
Encompass_Preferred
Chemical. CAS: 177931-17-8. Formula: C20H20O6. MW: 356.4. Isolated from Saururus chinensis. Hepatoprotective antioxidant. Anti-inflammatory. Suppresses NF-kB p65 activity. Neuroprotective. Apoptosis inhibitor. Bone resorption/osteoclastogenesis inhibitor. Reduces ROS formation. Antiseptic. AMP-activated kinase (AMPK) activator. Akt phosphorylation inhibitor.
Catalog No. 50-205-3377