Learn More
Surgical Design™ Sauerbruch Bone Rongeur
Catalog No. 21124469 Shop All Surgical Design Products
Change view
Click to view available options
Blade Style:
Straight
Catalog No. Blade Style
21-124-469 Straight
1 options
Catalog No. 21-124-469 Supplier Surgical Design™ Supplier No. 111274
Add to Cart
Add to Cart
Sauerbruch bone rongeur delivers exceptional strength for large bone removal with a 12 in. double-action stainless-steel design for powerful control.
  • Constructed from medical-grade stainless steel for outstanding durability and corrosion resistance
  • 12-in. length provides maximum reach and leverage for deep or large areas
  • Double-action mechanism multiplies cutting force for efficient bone removal with minimal effort
  • Heavy-duty jaws designed for powerful, clean resection of dense or cortical bone
  • Precision-ground edges ensure smooth, consistent performance
  • Ergonomic handle improves control and reduces hand fatigue
  • Balanced weight distribution enhances accuracy and stability in heavy use
  • Commonly used in forensic, autopsy, and pathology dissection
  • Suitable for trauma, reconstructive, and veterinary bone trimming procedures
  • Excellent for teaching and research environments requiring high-performance instruments
Specifications
Product Type Bone-cutting Forceps
Blade Material Stainless Steel
Blade Style Straight
Length (English) 12 in.
Length (Metric) 30.50 cm
Length (Metric) Blade 20 mm
Material Stainless Steel