Learn More
CPC Scientific Glp-Gly-Pro-Pro-Ile-Ser-Ile-Asp-Leu-Ser-Leu-Glu-Leu-Leu-Arg-Lys-Met-Ile-Glu-Ile-Glu-Lys-Gln-Glu-Lys-Glu-Lys-Gln-Gln-Ala-Ala-Asn-Asn-Arg-Leu-Leu-Leu-Asp-Thr-Ile-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CRFS006B
Encompass
Active polypeptide isolated from the skin of the neo-tropical frog Phyllomedusa Sauvagei that presents a number of pharmacological actions on diuresis, the cardiovascular system and the endocrine glands.ONE-LETTER SEQUENCE: Glp-GPPISIDLSLELLRKMIEIEKQEKEKQQAANNRLLLDTI-NH2MOLECULAR FORMULA: C202H346N56O63S1MOLECULAR WEIGHT:4599.38STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [74434-59-6], SYNONYMS: SauvagineRESEARCH AREA: HormonalREFERENCES: P.C. Montecucchi et al., Intl. J. Pept. Protein Res., 18, 113 (1981)
Catalog No. 50-210-9170