Learn More
Cayman Chemical SauvagIntrIfluoroACTAte500ug
SDP
Catalog No. 502950124
Encompass_Preferred
A neuropeptide hormone; reduces systemic blood pressure in normotensive rats at 0.5 UG/kg, s.c.; inhibits anterior pituitary release of prolactin, TSH, and growth hormone in rats; increases plasma ACTH levels in rats
Catalog No. 50-295-0124 Supplier Cayman Chemical Supplier No. 34442500 UG
A neuropeptide hormone; reduces systemic blood pressure in normotensive rats at 0.5 UG/kg, s.c.; inhibits anterior pituitary release of prolactin, TSH, and growth hormone in rats; increases plasma ACTH levels in rats