Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Savant™ SpeedVac™ DNA 130 Integrated Vacuum Concentrator System
GSA_VA

Catalog No. 13875331 Shop All Thermo Scientific Products
Change view
Click to view available options
Electrical Requirements:
115 V 60 Hz
230 V 50 Hz
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrical Requirements
13-875-331 115 V 60 Hz
13-875-332 230 V 50 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-875-331 Supplier Thermo Scientific™ Supplier No. DNA130115
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Save time and money with these Concentrator Kits. Everything you need for quick drying and concentrating of small sample volumes

Designed for fast, efficient concentration of DNA and RNA applications, the Thermo Scientific™ Savant SpeedVac™ DNA130 is a compact, integrated vacuum concentrator. SpeedVac™ DNA130 vacuum concentrator will support sample preparations with ease and efficiency.

  • Compact, easy to use design: smallest integrated benchtop unit saves lab bench space.
  • Corrosion-resistant, PTFE coated chamber offers years of service
  • Built-in, oil free vacuum pump offers maintenance-free operation
  • Includes a rotor that holds 36 x 1.5-2.0 ml tubes (RD36)

Operating Features:

  • Consistent performance: designed to ensure consistent drying times and reproducible results
  • User-friendly interface
  • Three user defined programs that can be modified and saved
  • Temperature set point: ambient, 35°C to 65°C in 5°C increments
  • Heat Time setting: from 1m to 9h and 59m and continuous
  • Run Time setting: from 1m to 9h and 59m
  • Audible alarm to alert user when run is complete

Sample Protection

  • Dry at ambient temperatures or using low heat to avoid sample denaturing
  • Oil-free diaphragm pump provides gentle transition from atmosphere to ultimate vacuum setting with no solvent bumping

Safety Features

  • PTFE-coated chamber resists chemicals and cleans easily
  • Latched lid and lid lock indicator ensure user safety during operation
  • Lid stay mechanism to secure lid while it‘s open, allowing user to have both hands free for operation
  • Unit and pump are virtually maintenance-free
  • External post trap to neutralize ammonium hydroxide vapors
  • Optional chemical traps and cartridges to trap radioactive chemicals to minimize concerns in handling hazaRDO™us fumes and chemicals.

Solvents and Solvent Combinations:

  • Water
  • PCR buffers (aqueous)
  • Ethanol and water
  • Ammonium Hydroxide (will require an ammonia post-trap and neutralizing solution accessory)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Temperature Range (Metric) Ambient to 65°C
Chamber Material PTFE-coated Aluminum
Description SpeedVac DNA130 Vacuum Concentrator System
Electrical Requirements 115 V 60 Hz
Amperage 10 A
Plug Type NEMA 5-15P
Weight (English) 86 lb.
Weight (Metric) 39 kg
Dimensions (L x W x H) 11.5 x 25 x 12 in. (29 x 64 x 31 cm)
Certifications UL Listed to US and Canadian requirements. CE marked
Max. Vacuum <10 Torr (13 mbar, 1.3k Pa)
Holds Accommodates 36 x 1.5 mL Tubes, 72 x 0.5 mL Tubes and 2 Microwell Plates
Includes RD36 rotor (36x 1.5-2.0 ml tubes)
Shipping Weight (English) 105 lb.
Voltage 115 V
Shipping Weight (Metric) 48 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.