Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits

Catalog No. 13875358
Change view
Click to view available options
Model:
SPD1030
SPD2030
Includes:
Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor
Diaphragm Pump, Cold Trap, Vacuum Gauge
Diaphragm Pump, Cold Trap, Vacuum Gauge, RH40-11 Rotor
Vacuum Concentrator
Electrical Requirements:
115 V 60 Hz
220 V 60 Hz
230 V 50 Hz
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes Electrical Requirements
13-875-358 SPD2030 Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor 230 V 50 Hz
13-875-339 SPD1030 Vacuum Concentrator 115 V 60 Hz
13-875-340 SPD1030 Vacuum Concentrator 230 V 50 Hz
13-875-359 SPD1030 Diaphragm Pump, Cold Trap, Vacuum Gauge 115 V 60 Hz
13-875-360 SPD1030 Diaphragm Pump, Cold Trap, Vacuum Gauge, RH40-11 Rotor 230 V 50 Hz
13-875-355 SPD1030 Diaphragm Pump, Cold Trap, Vacuum Gauge, RH40-11 Rotor 115 V 60 Hz
13-875-356 SPD1030 Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor 230 V 50 Hz
13-875-347 SPD2030 Diaphragm Pump, Cold Trap, Vacuum Gauge 220 V 60 Hz
13-875-348 SPD2030 Diaphragm Pump, Cold Trap, Vacuum Gauge 230 V 50 Hz
13-875-361 SPD2030 Diaphragm Pump, Cold Trap, Vacuum Gauge 220 V 60 Hz
13-875-362 SPD2030 Diaphragm Pump, Cold Trap, Vacuum Gauge 230 V 50 Hz
13-875-357 SPD2030 Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor 220 V 60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-875-358 Supplier Thermo Scientific™ Supplier No. SPD2030P1230
Only null left
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Get all the necessary components for HPLC fractions with water/0.1% trifluoroacetic acid and reversed-phase SPE eluants applications with these kits.

Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant SPD1030 and SPD2030 Integrated SpeedVac™ Systems and kits. Fully integrated for fast installation, programming. Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac SPD1030 System is designed for medium capacity (i.e.,120 x 1.5 mL tubes, other formats). SpeedVac SPD2030 System allows higher capacity (i.e.,200 x 1.5 mL tubes, other formats).

Easy to Use:

  • Plug and play installation:. Includes a concentrator, oil-free vacuum pump and cold trap all integrated in one unit. No tubing connections needed.
  • Intuitive and user friendly program digital display which allows use of three preset programs and ability to create three customizable programs to set heat, run time, ramp rate and vacuum level for efficient operation.
  • Wide variety of rotors to support sample format from micro tubes, vials and microplates to 250-ml flasks
  • Audible alarm to alert user when operation is finished

Robust, Low-Maintenance Design:

  • Oil-free vacuum pump is virtually maintenance free
  • PTFE coated chamber protects the chamber from corrosion, while making cleaning easy

Enhanced Evaporation:

  • Faster sample drying provided with radiant cover
  • Vacuum ramping and pressure controls eliminate sample bumping to maintain sample integrity
  • Reduced length vapor path from concentrator to refrigerated vapor trap accelerates evaporation
  • Rotors fit a wide variety of sample formats such as tubes, flasks, and shallow-well and deep-well microplates

Solvent and Solvent Combinations:

  • Acetonitrile
  • Ethanol
  • Methanol
  • Methylene Chloride
  • Water
  • Other organic solvents

SPD1030A and SPD2030A includes:

  • Integrated SpeedVac system
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD1030P1 Kit includes:

  • Fully Integrated SpeedVac system
  • 40 × 1.5mL rotor (RH40-11)
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD2030P1 kit

  • Integrated SpeedVac system
  • 200 × 1.5mL rotor (RH200-12)
  • CryoCool™ 1L heat transfer fluid
  • Glass condensation Flask (GCF400)
TRUSTED_SUSTAINABILITY

Specifications

Model SPD2030
Includes Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor
Shipping Weight (English) 260 lb.
Certifications UL Listed to US and Canadian requirements. CE marked
Max. Vacuum <10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (Metric) 136 kg
Holds Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Temperature Range (Metric) Ambient to 80°C
Chamber Material PTFE-coated Aluminum
Description SPD2030 Integrated System Kit
Electrical Requirements 230 V 50 Hz
Amperage 8 A
Weight (Metric) 98 kg
Dimensions (L x W x H) 31 x 28 x 19 in. (79 x 71 x42 cm)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.