Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Refrigerated Vapor Trap Series

Catalog No. 13875350 Shop All Thermo Scientific Products
Change view
Click to view available options
Electrical Requirements:
115 V 60 Hz
230 V 50 Hz
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrical Requirements
13-875-350 230 V 50 Hz
13-875-349 115 V 60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-875-350 Supplier Thermo Scientific™ Supplier No. RVT450230
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Condense and trap a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps.

Condenses and removes a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps (RVT). The RVT safely recovers the solvent and reduces emissions of hazardous chemicals in the environment.

Solvents are removed from samples during vacuum evaporation with a SpeedVac™ Concentrator which are then collected in a CFC-free Refrigerated Vapor Trap (RVT).   Solvent vapors pass into a Glass Condensation Flask and condense on its walls. This prevents the vapors from reaching and potentially damaging the oil-sealed vacuum pump. 

  • Rapid cooling time minimizes waiting period
  • Easy to remove glass flask for quick solvent changes
  • Rapidly cools to operating temperature
  • Easy to use glass condensation flask allows for the collection of liquid into a self-contained jar for easy draining
  • Sealed refrigeration system eliminates consumable coolants such as dry ice and acetone
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance UL Listed to US and Canadian requirements. CE marked
Description Refrigerated Vapor Cold Trap RVT450
Type Refrigerated Vapor Trap
Electrical Requirements 230 V 50 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.