Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 Large Capacity Vacuum Concentrator and Kits with Installation PROMO
Catalog No. 13875375IPM Shop All Thermo Scientific Products
Change view
Click to view available options
Electrical Requirements:
115 V, 60 Hz
Catalog No. Electrical Requirements
13-875-375IPM 115 V, 60 Hz
1 options
Catalog No. 13-875-375IPM Supplier Thermo Scientific™ Supplier No. SPD300P1115INSPM
Add to Cart
Add to Cart
Thermo Scientific™ SpeedVac™ Modular Vacuum Concentrators are designed to vaporate strong acids, bases and aggressive organic solvents
  • Approved for combinatorial chemistry
  • Incorporate an intuitive programmable user display for enhanced usability.
TRUSTED_SUSTAINABILITY
Specifications
Certifications UL Listed to US and Canadian requirements, CE marked
Electrical Requirements 115 V, 60 Hz