Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 Large Capacity Vacuum Concentrator and Kits with Installation
Catalog No. 13875375INS Shop All Thermo Scientific Products
Change view
Click to view available options
Certifications:
UL Listed to US and Canadian requirements. CE marked
Catalog No. Certifications
13-875-375INS UL Listed to US and Canadian requirements. CE marked
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-875-375INS Supplier Thermo Scientific™ Supplier No. SPD300P1115INS
Get Up to 55% Off Lab Equipment & Instruments
View all promotions
Add to Cart
Add to Cart
Request a Quote
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Thermo Scientific™ SpeedVac™ Modular Vacuum Concentrators are designed to vaporate strong acids, bases and aggressive organic solvents
  • Approved for combinatorial chemistry
  • Incorporate an intuitive programmable user display for enhanced usability.
TRUSTED_SUSTAINABILITY
Specifications
Description SpeedVac SPD300DDA Vacuum Concentrator Kit
Electrical Requirements 115 V 60 Hz
Certifications UL Listed to US and Canadian requirements. CE marked