Learn More
Enzo Life Sciences SAVIEW PLUS HRP reagent (150tests)
SDP
Supplier:  Enzo Life Sciences ENZACC1020150
Encompass_Preferred
SAVIEW PLUS HRP reagent is a ready-to-use streptavidin-based nanopolymer detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the SAVIEW PLUS HRP reagent can be used for in situ hybridization detection of biotinylated DNA/RNA probes. Long Term Storage: +4°C. Applications: IHC ISH (in situ hybridization).
Catalog No. 50-201-3354