Learn More
Selleck Chemical LLC Savolitinib(AZD6094) 2mg 1313725-88-0 HMPL-504
SDP
Catalog No. 501948080
Encompass_Preferred
Savolitinib (volitinib, AZD6094, HMPL-504) is a novel, potent, and selective MET inhibitor currently in clinical development in various indications, including PRCC. The IC50 values of this compound for c-Met and p-Met are 5 nM and 3 nM, respectively. It shows exquisite selectivity for c-Met over 274 kinase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8080 Supplier Selleck Chemical LLC Supplier No. S76742MG
Savolitinib (volitinib, AZD6094, HMPL-504) is a novel, potent, and selective MET inhibitor currently in clinical development in various indications, including PRCC. The IC50 values of this compound for c-Met and p-Met are 5 nM and 3 nM, respectively. It shows exquisite selectivity for c-Met over 274 kinase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.