Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SAvPhire Monomeric Streptavidin Alkaline Phosphatase Conjugate

Catalog No. SAE010910MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SAE010910MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SAE010910MI Supplier MilliporeSigma™ Supplier No. SAE0109100UG
Only null left
Add to Cart
Add to Cart

High-affinity recombinant variant of SA minimizing risk of aggregation, precipitation, and cross linking in biotinylated molecules

  • High-affinity recombinant variant of SA that minimizes the risk of aggregation, precipitation, and crosslinking in pre-incubation with biotinylated molecules
  • Offers easier access to the desired targets, tested for use in IHC (Paraffin)
  • Applications:
    • Immunohistochemistry (Paraffin)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 1.0 mg/mL
    Molecular Weight (g/mol) 15.5 kDa
    Quantity 100 μg
    Content And Storage Stable for 1 year (at 2°C to 8°C from date of receipt)
    Species Predicted to react with all species
    Buffer 0.05 M Tris, 0.15 M NaCl, 1 mM MgCl2, 0.2% BSA, pH 8.0 with 0.1% sodium azide with 10% glycerol
    Host Species E. coli

    For research purposes only

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.