Learn More
MilliporeSigma™ SAvPhire Monomeric Streptavidin Peroxidase Conjugate
Shop All MilliporeSigma ProductsDescription
- Minimizes risk of aggregation, precipitation, and crosslinking in pre-incubation with biotinylated molecules
- Offers easier access to desired targets
- Tested for use in IHC (Paraffin) and western blotting
Specifications
Specifications
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Molecular Weight (g/mol) | 15.5 kDa for Savphire monomeric streptavidin (with an N-terminal His-tag and a C-terminal FLAG tag) |
| Quantity | 100 μg |
| Content And Storage | Stable for 1 year (at 2°C to 8°C from date of receipt) |
| Species | Predicted to react with all species |
| Buffer | 0.15 M PBS, 0.5 % BSA, 0.4% Bronidox, pH 7.2 without azide with 10% glycerol |
| Host Species | E. coli |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.