Learn More
IMEB, Inc Pathology Bone Band Saw Optional Saw Blade with Fine Teeth
SDP
Catalog No. NC0907447
Encompass
Bone Band Saw Blade With Fine Teeth. Toothed Blade .138" wide X .025" X 24tpi for the IMEB, Inc. Bone Band Saw which is sold separately. IMEB Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.
Catalog No. NC0907447 Supplier IMEB, Inc Supplier No. SFF532
Bone Band Saw Blade With Fine Teeth. Toothed Blade .138" wide X .025" X 24tpi for the IMEB, Inc. Bone Band Saw which is sold separately. IMEB Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.