Learn More
IMEB, Inc Pathology Bone Band Saw Optional Saw Blade with Heavy Duty Teeth
SDP
Catalog No. NC0907444
Encompass
Saw Blade with Heavy Duty Teeth Toothed Blade is 180" wide X .031" X 14tpi. IMEB Bone Band Saw sold separately. The IMEB, Inc. Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw is only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.
Catalog No. NC0907444 Supplier IMEB, Inc Supplier No. SH303
Saw Blade with Heavy Duty Teeth Toothed Blade is 180" wide X .031" X 14tpi. IMEB Bone Band Saw sold separately. The IMEB, Inc. Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw is only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.