Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore SAX LC Analytical Column, 10 m, 4.650 mm, PEEK
Catalog No. NC2815230
Encompass_Preferred
BioCore SAX is a family of high-performance, strong anion-exchange columns designed for separating charged variants in low pI proteins, such as fusion proteins. Its column technology involves the creation of a hydrophilic quaternary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC2815230 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B61110000004605P
BioCore SAX is a family of high-performance, strong anion-exchange columns designed for separating charged variants in low pI proteins, such as fusion proteins. Its column technology involves the creation of a hydrophilic quaternary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.