Learn More
NANOMICRO TECHNOLOGIES INC BioCore SAX LC Analytical Column, 5um, 4.6x50 mm, PEEK
Supplier:  NANOMICRO TECHNOLOGIES INC B61105000004605P
Encompass_Preferred
BioCore SAX is a family of high-performance, strong anion-exchange columns designed for separating charged variants in low pI proteins, such as fusion proteins. Its column technology involves the creation of a hydrophilic quaternary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC2815245