Learn More
Analytical Sales & Services SAX SPE 96-Well Plate, 1.0mL/100mg Bed
Catalog No. 5000806224
Encompass
Silica Based Strong Anion Exchange SPE, 1mL 96 Well Filter Plate, 100mg per well
Catalog No. 50-008-06224 Supplier Analytical Sales & Services Supplier No. 9610SAX100
Silica Based Strong Anion Exchange SPE, 1mL 96 Well Filter Plate, 100mg per well