Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Analytical Sales & Services SAX SPE 96-Well Plate, 2.0mL/25mg Bed

Catalog No. 5000804265
Encompass

Strong Anion Exchange SPE, 2mL 96 Well Filter Plate, 25mg per well

Catalog No. 50-008-04265 Supplier Analytical Sales & Services Supplier No. 9620SAX25
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strong Anion Exchange SPE, 2mL 96 Well Filter Plate, 25mg per well

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.