Learn More
NANOMICRO TECHNOLOGIES INC BioCore SAX LC Preparative Column, 5um, 21.2x250 mm, Stainless Steel
Catalog No. NC2815246
Encompass_Preferred
BioCore SAX is a family of high-performance, strong anion-exchange columns designed for separating charged variants in low pI proteins, such as fusion proteins. Its column technology involves the creation of a hydrophilic quaternary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC2815246 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B61105000021225S
Add to Cart
Add to Cart
Request a Quote
BioCore SAX is a family of high-performance, strong anion-exchange columns designed for separating charged variants in low pI proteins, such as fusion proteins. Its column technology involves the creation of a hydrophilic quaternary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.