Learn More
Medchemexpress LLC HY-10285 10mg Medchemexpress, Saxagliptin CAS:361442-04-8 Purity:>98%
SDP
Catalog No. 501968001
Encompass_Preferred
Medchemexpress, HY-10285 10mg Saxagliptin CAS:361442-04-8 Saxagliptin is an orally active dipeptidyl peptidase-4 (DPP4) inhibitor that helps control blood sugar levels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8001 Supplier Medchemexpress LLC Supplier No. HY1028510MG
Medchemexpress, HY-10285 10mg Saxagliptin CAS:361442-04-8 Saxagliptin is an orally active dipeptidyl peptidase-4 (DPP4) inhibitor that helps control blood sugar levels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.