Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saxagliptin hydrate | 945667-22-1 | MFCD26131254 | ≥99.0% | 333.43 | C18H27N3O3 | 100 MG
SDP

Catalog No. 5000245251
Encompass_Preferred

Saxagliptin hydrate is a potent, selective, reversible, competitive, orally active dipeptidyl peptidase-4 (DPP-4) inhibitor used for research into type 2 diabetes and DPP-4 biology. It is offered in solid and solution forms across multiple pack sizes and is intended for research use only.

  • Research use only; not for human or clinical use.
  • CAS number 945667-22-1.
  • Purity typically ≥99.0% (data sheet reports 98.13%).
  • Molecular weight 333.43 and formula C18H27N3O3.
  • Available as solids (5-100 mg and bulk) and 10 mM solutions in DMSO.
  • Supplied quantities suitable for assay development and preclinical research.

Catalog No. 50-002-45251 Supplier Medchemexpress LLC Supplier No. HY10285A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saxagliptin hydrate is a potent, selective, reversible, competitive, orally active dipeptidyl peptidase-4 (DPP-4) inhibitor used for research into type 2 diabetes and DPP-4 biology. It is offered in solid and solution forms across multiple pack sizes and is intended for research use only.

  • Research use only; not for human or clinical use.
  • CAS number 945667-22-1.
  • Purity typically ≥99.0% (data sheet reports 98.13%).
  • Molecular weight 333.43 and formula C18H27N3O3.
  • Available as solids (5-100 mg and bulk) and 10 mM solutions in DMSO.
  • Supplied quantities suitable for assay development and preclinical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.