Learn More
Selleck Chemical LLC Saxagliptin hydrochloride E6005-25MG
SDP
Catalog No. 5000041414
Encompass_Preferred
Saxagliptin hydrochloride (BMS-477118 hydrochloride) is a potent selective reversible competitive and orally active inhibitor of dipeptidyl peptidase-4 (DPP-4) It exhibits 400-950 fold selective inhibition of DPP-4 than DPP-8 Saxagliptin hydrochloride is used in the research of type 2 diabetes mellitus research
Catalog No. 50-000-41414 Supplier Selleck Chemical LLC Supplier No. E600525MG
Saxagliptin hydrochloride (BMS-477118 hydrochloride) is a potent selective reversible competitive and orally active inhibitor of dipeptidyl peptidase-4 (DPP-4) It exhibits 400-950 fold selective inhibition of DPP-4 than DPP-8 Saxagliptin hydrochloride is used in the research of type 2 diabetes mellitus research