Learn More
Medchemexpress LLC Saxagliptin (hydrochloride) | 709031-78-7 | 99.7% | 351.87 | 25 MG
SDP
Catalog No. 5000393053
Encompass_Preferred
Saxagliptin hydrochloride is a potent, selective, reversible, competitive, and orally active dipeptidyl peptidase-4 (DPP-4) inhibitor with a Ki of 0.6-1.3 nM. This compound has potential for research into type 2 diabetes mellitus.
  • Potent inhibitor of dipeptidyl peptidase-4 (DPP-4).
  • Selective and reversible action.
  • Orally active compound.
  • Increases β-cell proliferation.
  • Increases p-AKT and active β-catenin protein levels.
  • Increases c-myc and cyclin D1 protein expression.
  • Prevents degradation of glucagon-like peptide-1.
  • Increases secretion of insulin.
  • Decreases secretion of glucagon.
Catalog No. 50-003-93053 Supplier Medchemexpress LLC Supplier No. 25MGHY16448
Saxagliptin hydrochloride is a potent, selective, reversible, competitive, and orally active dipeptidyl peptidase-4 (DPP-4) inhibitor with a Ki of 0.6-1.3 nM. This compound has potential for research into type 2 diabetes mellitus.
  • Potent inhibitor of dipeptidyl peptidase-4 (DPP-4).
  • Selective and reversible action.
  • Orally active compound.
  • Increases β-cell proliferation.
  • Increases p-AKT and active β-catenin protein levels.
  • Increases c-myc and cyclin D1 protein expression.
  • Prevents degradation of glucagon-like peptide-1.
  • Increases secretion of insulin.
  • Decreases secretion of glucagon.