Learn More
Medchemexpress LLC Saxagliptin hydrochloride | 709031-78-7 | 99.7% | 351.87 | 5 MG
SDP
Catalog No. 5000392447
Encompass_Preferred
Saxagliptin hydrochloride is a potent, selective, reversible, competitive, and orally active dipeptidyl peptidase-4 (DPP-4) inhibitor. It has the potential for type 2 diabetes mellitus research.
  • Potent, selective, reversible, competitive, and orally active DPP-4 inhibitor
  • Induces β-cell proliferation
  • Increases p-AKT and active β-catenin protein levels
  • Prevents glucagon-like peptide-1 degradation
  • Increases insulin secretion and decreases glucagon secretion
Catalog No. 50-003-92447 Supplier Medchemexpress LLC Supplier No. HY164485MG
Saxagliptin hydrochloride is a potent, selective, reversible, competitive, and orally active dipeptidyl peptidase-4 (DPP-4) inhibitor. It has the potential for type 2 diabetes mellitus research.
  • Potent, selective, reversible, competitive, and orally active DPP-4 inhibitor
  • Induces β-cell proliferation
  • Increases p-AKT and active β-catenin protein levels
  • Prevents glucagon-like peptide-1 degradation
  • Increases insulin secretion and decreases glucagon secretion