Learn More
Medchemexpress LLC Saxagliptin hydrochloride | 709031-78-7 | 99.7% | 351.87 | 50 MG
SDP
Catalog No. 5000392425
Encompass_Preferred
Saxagliptin hydrochloride is a potent, selective, reversible, competitive, and orally active dipeptidyl peptidase-4 (DPP-4) inhibitor with a Ki of 0.6-1.3 nM. It has potential for research into type 2 diabetes mellitus. It has been shown to induce β-cell proliferation, increase p-AKT and active β-catenin protein levels, and increase insulin secretion.
  • Potent, selective, reversible, competitive, and orally active
  • Inhibits dipeptidyl peptidase-4 (DPP-4) with a Ki of 0.6-1.3 nM
  • Potential for type 2 diabetes mellitus research
  • Induces β-cell proliferation
  • Increases p-AKT and active β-catenin protein levels
  • Increases insulin secretion and decreases glucagon secretion
Catalog No. 50-003-92425 Supplier Medchemexpress LLC Supplier No. HY1644850MG
Saxagliptin hydrochloride is a potent, selective, reversible, competitive, and orally active dipeptidyl peptidase-4 (DPP-4) inhibitor with a Ki of 0.6-1.3 nM. It has potential for research into type 2 diabetes mellitus. It has been shown to induce β-cell proliferation, increase p-AKT and active β-catenin protein levels, and increase insulin secretion.
  • Potent, selective, reversible, competitive, and orally active
  • Inhibits dipeptidyl peptidase-4 (DPP-4) with a Ki of 0.6-1.3 nM
  • Potential for type 2 diabetes mellitus research
  • Induces β-cell proliferation
  • Increases p-AKT and active β-catenin protein levels
  • Increases insulin secretion and decreases glucagon secretion