Learn More
Abraxis Saxitoxin (PSP) ELISA, Microtiter Plate, 96 tests, detection limit 0.015 ng/mL, approx 1 hour assay time
SDP
Supplier:  Abraxis 52255B
Encompass
Saxitoxin (PSP) ELISA, Microtiter Plate, 96 tests, detection limit 0.015 ng/mL, approx 1 hour assay time
Catalog No. NC1394972