Learn More
MSC DBI-SALA 3M Sayfline Cable Lifeline, Galvanized, 2-Worker, 1 ea
Catalog No. 5000674587
Encompass
The DBI-SALA Cable Horizontal Lifeline System is an all-metal, galvanized cable system engineered for fall protection in construction or general industry. This durable yet lightweight system is easily transportable between job sites.
  • Galvanized cable lifeline available in multiple lengths
  • Metal energy absorbers limit lifeline tension forces, protecting structures and minimizing fall clearance
  • Built-in turnbuckle tensioner for system tensioning
  • All-metal design suitable for extended placement
  • Accommodates two 310 lb (140.6 kg) workers per span, up to 6 workers per system with multiple spans
  • RFID Wedge Grip termination device for quicker setup and modifications
  • Horizontal design allows free movement without needing new anchorage points
  • Meets OSHA regulations
Catalog No. 50-006-74587 Supplier MSC Supplier No. 58958570
The DBI-SALA Cable Horizontal Lifeline System is an all-metal, galvanized cable system engineered for fall protection in construction or general industry. This durable yet lightweight system is easily transportable between job sites.
  • Galvanized cable lifeline available in multiple lengths
  • Metal energy absorbers limit lifeline tension forces, protecting structures and minimizing fall clearance
  • Built-in turnbuckle tensioner for system tensioning
  • All-metal design suitable for extended placement
  • Accommodates two 310 lb (140.6 kg) workers per span, up to 6 workers per system with multiple spans
  • RFID Wedge Grip termination device for quicker setup and modifications
  • Horizontal design allows free movement without needing new anchorage points
  • Meets OSHA regulations