Learn More
As One International Inc 10x Standard Buffer, 10 x 1.5 ml
Catalog No. 501655971
Encompass_Preferred
Standard Buffer is a traditional potassium buffer and has high specificity. However, optimization of primer annealing temperatures and magnesium concentration is required.
Catalog No. 50-165-5971 Supplier As One International Inc Supplier No. AO302110
Standard Buffer is a traditional potassium buffer and has high specificity. However, optimization of primer annealing temperatures and magnesium concentration is required.