Learn More
Selleck Chemical LLC SB 200646 S2649-5mg
SDP
Catalog No. 503110226
Encompass_Preferred
SB 200646 (SB 200646A) is a potent selective and oral-active antagonist of 5-HT2B/2C over 5-HT2A receptor with 50 fold selectivity The pKi for rat 5-HT2C receptor rat 5-HT2B receptor and rat 5-HT2A receptor are 6 9 7 5 and 5 2 respectively SB 200646 has electrophysiological and anxiolytic properties in vivo
Catalog No. 50-311-0226 Supplier Selleck Chemical LLC Supplier No. S26495MG
SB 200646 (SB 200646A) is a potent selective and oral-active antagonist of 5-HT2B/2C over 5-HT2A receptor with 50 fold selectivity The pKi for rat 5-HT2C receptor rat 5-HT2B receptor and rat 5-HT2A receptor are 6 9 7 5 and 5 2 respectively SB 200646 has electrophysiological and anxiolytic properties in vivo