Learn More
Medchemexpress LLC HY-10295 100mg Medchemexpress, SB 202190 CAS:152121-30-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10295100MG
Encompass_Preferred
Medchemexpress, HY-10295 100mg SB 202190 CAS:152121-30-7 SB 202190 is a selective p38 MAP kinase inhibitor with IC50s of 50 nM and 100 nM for p38α and p38β2, respectively. SB 202190 binds to the ATP pocket of the active recombinant human p38 kinase with a Kd of 38 nM. SB 202190 has anti-cancer activity and rescued memory deficits[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8718