Learn More
Medchemexpress LLC HY-10295 200mg Medchemexpress, SB 202190 CAS:152121-30-7 Purity:98%
SDP
Catalog No. 501637104
Encompass_Preferred
Medchemexpress,HY-10295 200mg SB 202190 CAS:152121-30-7 Formula:C20H14FN3O SB 202190 is a cell-permeable p38 MAP kinase inhibitor with IC50s of 50 nM and 100 nM for p38 and p38β2, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7104 Supplier Medchemexpress LLC Supplier No. HY10295200MG
Medchemexpress,HY-10295 200mg SB 202190 CAS:152121-30-7 Formula:C20H14FN3O SB 202190 is a cell-permeable p38 MAP kinase inhibitor with IC50s of 50 nM and 100 nM for p38 and p38β2, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.