Learn More
Selleck Chemical LLC SB202190 (FHPI) 25mg 152121-30-7
SDP
Catalog No. 507979
Encompass_Preferred
SB202190 (FHPI) is a potent p38 MAPK inhibitor targeting p38?/? with IC50 of 50 nM/100 nM in cell-free assays, sometimes used instead of SB 203580 to investigate potential roles for SAPK2a/p38 in vivo. SB202190 inhibits endothelial cell apoptosis via induction of autophagy and heme oxygenase-1. SB202190 significantly suppresses Erastin?dependent ferroptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-797-9 Supplier Selleck Chemical LLC Supplier No. S107725MG
SB202190 (FHPI) is a potent p38 MAPK inhibitor targeting p38?/? with IC50 of 50 nM/100 nM in cell-free assays, sometimes used instead of SB 203580 to investigate potential roles for SAPK2a/p38 in vivo. SB202190 inhibits endothelial cell apoptosis via induction of autophagy and heme oxygenase-1. SB202190 significantly suppresses Erastin?dependent ferroptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.