Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB 202190 hydrochloride | 350228-36-3 | MFCD17215958 | ≥98.0% | 367.8 g/mol | C20H15ClFN3O | 100 MG
SDP

Supplier:  Medchemexpress LLC HY10295A100MG

Encompass_Preferred

SB 202190 hydrochloride is a selective inhibitor of p38 mitogen-activated protein kinases used in research to probe p38α/β signaling pathways. It is supplied as the monohydrochloride salt in crystalline form and is intended for research use only.

  • Selective p38α and p38β inhibitor with IC50 values of 50 nM and 100 nM.
  • High affinity binding to the p38 ATP pocket (Kd ≈ 38 nM).
  • Molecular weight approximately 367.8 g/mol.
  • Empirical formula: C20H15ClFN3O (hydrochloride salt).
  • Physical form: crystalline solid; storage at -20°C; in solution store at -80°C for long-term.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-48985


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.