Learn More
Medchemexpress LLC SB 202190 hydrochloride | 350228-36-3 | MFCD17215958 | 99.7% | 367.80 g·mol⁻1 | C20H15ClFN3O | 50 MG
SDP
Catalog No. 5000246593
Encompass_Preferred
SB 202190 hydrochloride is the hydrochloride salt of a selective p38 mitogen-activated protein kinase (MAPK) inhibitor used in biochemical and cellular research to probe p38 signaling pathways. It is supplied as a solid and as a ready-to-use DMSO solution, and is characterized by high purity and defined storage recommendations.
  • Selective inhibitor of p38 MAPK isoforms.
  • High purity suitable for research applications.
  • Available as solid and as a 10 mM solution in DMSO.
  • Light yellow solid appearance for easy visual inspection.
  • Requires low-temperature, sealed storage to preserve stability.
Catalog No. 50-002-46593 Supplier Medchemexpress LLC Supplier No. HY10295A50MG
SB 202190 hydrochloride is the hydrochloride salt of a selective p38 mitogen-activated protein kinase (MAPK) inhibitor used in biochemical and cellular research to probe p38 signaling pathways. It is supplied as a solid and as a ready-to-use DMSO solution, and is characterized by high purity and defined storage recommendations.
  • Selective inhibitor of p38 MAPK isoforms.
  • High purity suitable for research applications.
  • Available as solid and as a 10 mM solution in DMSO.
  • Light yellow solid appearance for easy visual inspection.
  • Requires low-temperature, sealed storage to preserve stability.