Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB 202190 hydrochloride | 350228-36-3 | MFCD28391789 | 99.7% | 367.80 g·mol⁻¹ | C20H15ClF3NO | 1 ML
SDP

Catalog No. 5000246594
Encompass_Preferred

SB 202190 hydrochloride is a selective p38 MAP kinase inhibitor used in cell signaling and pharmacology research. It is commonly supplied in concentrated stock formats for convenient preparation of working solutions.

  • Selective p38 MAPK inhibitor; p38α IC50 ≈ 50 nM, p38β2 IC50 ≈ 100 nM.
  • Supplied in a 10 mM solution format (1 mL) for ready stock preparation.
  • Molecular weight 367.80 g·mol⁻¹; formula C20H15ClF3NO.
  • DMSO-soluble; may require sonication and warming (up to 60°C) for full dissolution.
  • Storage in solvent: -80°C (up to 6 months) or -20°C (up to 1 month); keep sealed and dry.
  • High purity (manufacturer reported ~99.7%) suitable for research applications.

Catalog No. 50-002-46594 Supplier Medchemexpress LLC Supplier No. HY10295A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB 202190 hydrochloride is a selective p38 MAP kinase inhibitor used in cell signaling and pharmacology research. It is commonly supplied in concentrated stock formats for convenient preparation of working solutions.

  • Selective p38 MAPK inhibitor; p38α IC50 ≈ 50 nM, p38β2 IC50 ≈ 100 nM.
  • Supplied in a 10 mM solution format (1 mL) for ready stock preparation.
  • Molecular weight 367.80 g·mol⁻¹; formula C20H15ClF3NO.
  • DMSO-soluble; may require sonication and warming (up to 60°C) for full dissolution.
  • Storage in solvent: -80°C (up to 6 months) or -20°C (up to 1 month); keep sealed and dry.
  • High purity (manufacturer reported ~99.7%) suitable for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.