Learn More
Medchemexpress LLC HY-10256 50mg Medchemexpress, SB 203580 CAS:152121-47-6 Purity:>98%
SDP
Catalog No. 502028713
Encompass_Preferred
Medchemexpress, HY-10256 50mg SB 203580 CAS:152121-47-6 SB 203580 (RWJ 64809) is a selective and ATP-competitive p38 MAPK inhibitor with IC50s of 50 nM and 500 nM for SAPK2a/p38 and SAPK2b/p38β2, respectively. SB 203580 inhibits LCK, GSK3β and PKBα with IC50s of 100-500-fold higher than that for SAPK2a/p38. SB 203580 does not disrupt JNK activity and is an autophagy and mitophagy activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8713 Supplier Medchemexpress LLC Supplier No. HY1025650MG
Medchemexpress, HY-10256 50mg SB 203580 CAS:152121-47-6 SB 203580 (RWJ 64809) is a selective and ATP-competitive p38 MAPK inhibitor with IC50s of 50 nM and 500 nM for SAPK2a/p38 and SAPK2b/p38β2, respectively. SB 203580 inhibits LCK, GSK3β and PKBα with IC50s of 100-500-fold higher than that for SAPK2a/p38. SB 203580 does not disrupt JNK activity and is an autophagy and mitophagy activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.