Learn More
Selleck Chemical LLC SB203580 25mg 152121-47-6 RWJ 64809, PB 203580
SDP
Catalog No. 507974
Encompass_Preferred
SB203580 (RWJ 64809, PB 203580) is a p38 MAPK inhibitor with IC50 of 0.3-0.5 ?M in THP-1 cells, 10-fold less sensitive to SAPK3(106T) and SAPK4(106T) and blocks PKB phosphorylation with IC50 of 3-5 ?M. SB203580 induces mitophagy and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-797-4 Supplier Selleck Chemical LLC Supplier No. S107625MG
SB203580 (RWJ 64809, PB 203580) is a p38 MAPK inhibitor with IC50 of 0.3-0.5 ?M in THP-1 cells, 10-fold less sensitive to SAPK3(106T) and SAPK4(106T) and blocks PKB phosphorylation with IC50 of 3-5 ?M. SB203580 induces mitophagy and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.